Stampa…

Montelukast sodium ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: MK-476, Sodium (R,E)-2-(1-(((1-(3-(2-(7-chloroquinolin-2-yl)vinyl)phenyl)-3-(2-(2-hydroxypropan-2-yl)phenyl)propyl)thio)methyl)cyclopropyl)acetate

M2340-200MG M2340-1G
TCIAM2340-200MGEA 28 EUR
TCIAM2340-200MG TCIAM2340-1G
Montelukast sodium ≥98.0% (by HPLC, titration analysis)
Montelukast sodium
Formula: C₃₅H₃₆ClNO₃S
Peso molecolare: 608,17 g/mol
MDL Number: MFCD00931431
CAS Number: 151767-02-1
Merck Index: 14,06258
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48b9f8c82a04c227d1ebb76b4d87cbda1a1d0dc2&vid=48b9f8c82a04c227d1ebb76b4d87cbda1a1d0dc2&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Specific rotation [a]20/D +95.0 to +105.0 deg (C=1、MeOH)
Water max. 7.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR