Stampa…
1-Methyl-1H-pyrazole-4-boronic acid pinacol ester ≥98.0% (by GC)
Fornitore: TCI EUROPE
Attenzione
Synonyms: 1-Methylpyrazole-4-boronic acid pinacol ester, 1-Methyl-4-pyrazole boronic acid pinacol ester, 1-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
M2339-1G M2339-5G
TCIAM2339-1GEA 50 EUR
TCIAM2339-1G TCIAM2339-5G
1-Methyl-1H-pyrazole-4-boronic acid pinacol ester ≥98.0% (by GC)
1-Methyl-1H-pyrazole-4-boronic acid pinacol ester
Formula: C₁₀H₁₇BN₂O₂
Peso molecolare: 208,07 g/mol
MDL Number: MFCD03789259
CAS Number: 761446-44-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f4f480b78608e6d456f6182111aadeecbaf7bed&vid=4f4f480b78608e6d456f6182111aadeecbaf7bed&prid=web&clid=SAPDAM
Visualizza i codici
Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 60.0 to 64.0 deg-C