Stampa…

N-Methoxycarbonylmaleimide ≥98.0% (by total nitrogen basis)

Fornitore: TCI EUROPE

Synonyms: Methyl 2,5-dioxo-2,5-dihydro-1H-pyrrole-1-carboxylate

M2333-1G M2333-5G
TCIAM2333-1GEA 42 EUR
TCIAM2333-1G TCIAM2333-5G
N-Methoxycarbonylmaleimide ≥98.0% (by total nitrogen basis)
N-Methoxycarbonylmaleimide
Formula: C₆H₅NO₄
Peso molecolare: 155,11 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00042755
CAS Number: 55750-48-6
EINECS: 259-792-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f1fe5adf860f3de891581e58db950e9b274cc90a&vid=f1fe5adf860f3de891581e58db950e9b274cc90a&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Dark green powder to crystaline
Purity (with Total Nitrogen) min. 98.0 %
Melting point 64.0 to 68.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR