Stampa…

Methyl hydrogen isophthalate ≥97.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Isophthalic acid monomethyl ester, 3-(Methoxycarbonyl)benzoic acid, Benzene-1,3-dicarboxylic acid monomethyl ester, mono-Methyl isophthalate

M2327-1G M2327-5G
TCIAM2327-1GEA 24 EUR
TCIAM2327-1G TCIAM2327-5G
Methyl hydrogen isophthalate ≥97.0% (by GC)
Methyl hydrogen isophthalate
Formula: C₉H₈O₄
Peso molecolare: 180,16 g/mol
MDL Number: MFCD00029972
CAS Number: 1877-71-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=91fc74587d788ec7552dfac3038809a0f9748e33&vid=91fc74587d788ec7552dfac3038809a0f9748e33&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 95.0 %
Melting point 192.0 to 196.0 deg-C (after drying)
Drying loss max. 4.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR