Methyl-4-aminonicotinate ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Methyl 4-aminonicotinate
M2303-1G
M2303-5G
TCIAM2303-1GEA
145
EUR
TCIAM2303-1G
TCIAM2303-5G
Methyl-4-aminonicotinate ≥98.0% (by GC, titration analysis)
Methyl-4-aminonicotinate
Formula:
C₇H₈N₂O₂ Peso molecolare: 152,15 g/mol Punto di fusione: 174°C |
MDL Number:
MFCD00955996 CAS Number: 16135-36-7 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...