Stampa…

Miglitol ≥98.0% (by GC, titration analysis)

Fornitore: TCI

Synonyms: Diastabol, BAY1099, Seibule, BAY-m1099, Glyset, (2R,3R,4R,5S)-1-(2-Hydroxyethyl)-2-(hydroxymethyl)piperidine-3,4,5-triol

M2302-5G M2302-1G
TCIAM2302-5GEA 290 EUR
TCIAM2302-5G TCIAM2302-1G
Miglitol ≥98.0% (by GC, titration analysis)
Miglitol
Formula: C₈H₁₇NO₅
Peso molecolare: 207.23 g/mol
MDL Number: MFCD00867240
CAS Number: 72432-03-2
EINECS: 276-661-6
Merck Index: 14,06187

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 144.0 to 148.0 deg-C
Specific rotation [a]20/D -7.0 to -9.0 deg (C=0.5、H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR