Stampa…

3-Amino-4-methylbenzotrifluoride ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

M2301-1G M2301-5G
TCIAM2301-1GEA 35 EUR
TCIAM2301-1G TCIAM2301-5G
3-Amino-4-methylbenzotrifluoride ≥98.0% (by GC, titration analysis)
3-Amino-4-methylbenzotrifluoride
Formula: C₈H₈F₃N
Punto di ebollizione: 74°C
MDL Number: MFCD00134580
CAS Number: 25449-96-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0e827baf55704eae4d47b7c816162dbeb736c6a5&vid=0e827baf55704eae4d47b7c816162dbeb736c6a5&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to lump
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 38.0 to 42.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR