Stampa…

Meropenem trihydrate ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI

M2279-1G M2279-5G
TCIAM2279-1GEA 78 EUR
TCIAM2279-1G TCIAM2279-5G
Meropenem trihydrate ≥98.0% (by HPLC, titration analysis)
Meropenem trihydrate
Formula: C₁₇H₃₁N₃O₈S
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD08600005
CAS Number: 119478-56-7
Merck Index: 14,05900

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 % (calcd.on anh.substance)
Specific rotation [a]20/D -17.0 to -21.0 deg (C=0.44, H2O)
Water 11.4 to 13.4 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR