Tacrolimus monohydrate ≥80.0% (by HPLC)
Fornitore: TCI
|
Pericolo
|
Synonyms:
FK 506, (3S,4R,5S,8R,9E,12S,14S,15R,16S,18R,19R,26aS)-5,6,8,11,12,13,14,15,16,17,18,19,24,25,26,26a-Hexadecahydro-5,19-dihydroxy-3-[(1E)-2-[(1R,3R,4R)-4-hydroxy-3-methoxycyclohexyl]-1-methylethenyl]-14,16-dimethoxy-4,10,12,18-tetramethyl-8-(2-propen-1-yl)-15,19-epoxy-3H-pyrido[2,1-c][1,4]oxaazacyclotricosine-1,7,20,21(4H,23H)-tetrone monohydrate
Formula:
C₄₄H₇₁NO₁₃ Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD11045918 CAS Number: 109581-93-3 UN: 2811 ADR: 6.1,III Merck Index: 14,09025 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 80.0 area% |
Melting point | 125.0 to 129.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...