Stampa…

N-Methylpentylamine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: N-Pentylmethylamine

M2231-1G M2231-5G
TCIAM2231-1GEA 43 EUR
TCIAM2231-1G TCIAM2231-5G
N-Methylpentylamine ≥98.0% (by GC, titration analysis)
N-Methylpentylamine
Formula: C₆H₁₅N
Peso molecolare: 101,19 g/mol
Punto di ebollizione: 118°C
Flash Pt: 12°C
MDL Number: MFCD00041354
CAS Number: 25419-06-1
EINECS: 246-966-9
UN: 2733
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1c2ae172a9593ae3ee93a9facd946f0a00f5c1bf&vid=1c2ae172a9593ae3ee93a9facd946f0a00f5c1bf&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR