1-Boc-isonipecotic acid methyl ester ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Methyl piperidine-4-carboxylate, N-BOC protected, 1-tert-Butyl 4-methyl piperidine-1,4-dicarboxylate
M2227-1G
M2227-5G
TCIAM2227-1GEA
24
EUR
TCIAM2227-1G
TCIAM2227-5G
1-Boc-isonipecotic acid methyl ester ≥98.0% (by GC)
1-Boc-isonipecotic acid methyl ester
Formula:
C₁₂H₂₁NO₄ Peso molecolare: 243,2994 g/mol |
MDL Number:
MFCD02183584 CAS Number: 124443-68-1 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to lump |
Purity (GC) | min. 98.0 % |
Melting point | 33.0 to 37.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...