Stampa…

Methyl (S)-(+)-2-pyrrolidone-5-carboxylate ≥98.0% (by GC)

Fornitore: TCI
Attenzione

Synonyms: Methyl (S)-(+)-2-oxopyrrolidine-5-carboxylate, H-Pyr-OMe

M2198-100G M2198-25G
TCIAM2198-100GEA 121 EUR
TCIAM2198-100G TCIAM2198-25G
Methyl (S)-(+)-2-pyrrolidone-5-carboxylate ≥98.0% (by GC)
Methyl (S)-(+)-2-pyrrolidone-5-carboxylate
Formula: C₆H₉NO₃
Peso molecolare: 143.14 g/mol
Punto di ebollizione: 135°C
MDL Number: MFCD00080931
CAS Number: 4931-66-2
EINECS: 225-567-3

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Specific rotation [a]20/D +9.0 to +12.0 deg (C=1, ethanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR