1-Methyl-4-[1-(4-piperidyl)-4-piperidyl]piperazine ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
1-Methyl-4-(1,4'-bipiperidin-4-yl)piperazine
M2197-25G
M2197-5G
TCIAM2197-25GEA
434
EUR
TCIAM2197-25G
TCIAM2197-5G
1-Methyl-4-[1-(4-piperidyl)-4-piperidyl]piperazine ≥98.0% (by GC, titration analysis)
1-Methyl-4-[1-(4-piperidyl)-4-piperidyl]piperazine
Formula:
C₁₅H₃₀N₄ Peso molecolare: 266,43 g/mol |
MDL Number:
MFCD20257913 CAS Number: 1629218-99-0 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 87.0 to 91.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...