N-Methylmorpholine-N-oxide ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
4-Methylmorpholine N-Oxide, 4-Methylmorpholine 4-oxide
M2192-5G
M2192-25G
TCIAM2192-5GEA
37
EUR
TCIAM2192-5G
TCIAM2192-25G
N-Methylmorpholine-N-oxide ≥98.0% (by titrimetric analysis)
N-Methylmorpholine-N-oxide
Formula:
C₅H₁₁NO₂ Peso molecolare: 117,15 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00005947 CAS Number: 7529-22-8 EINECS: 231-391-8 |
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...