Stampa…

Methylmaleic hydrazide ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3,6-Dihydroxy-4-methylpyridazine, 4-Methyl-3,6-pyridazinediol, 1,2-Dihydro-4-methyl-3,6-pyridazinedione, Methylmaleic Acid Hydrazide

M2182-25G M2182-5G
TCIAM2182-25GEA 151 EUR
TCIAM2182-25G TCIAM2182-5G
Methylmaleic hydrazide ≥98.0% (by GC)
Methylmaleic hydrazide
Formula: C₅H₆N₂O₂
Peso molecolare: 126,12 g/mol
Punto di fusione: 287°C
MDL Number: MFCD00067746
CAS Number: 5754-18-7
EINECS: 227-276-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR