Stampa…

Mirtazapine ≥98.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: 1,2,3,4,10,14b-Hexahydro-2-methylpyrazino[2,1-a]pyrido[2,3-c][2]benzazepine

M2151-1G M2151-100MG
TCIAM2151-1GEA 206 EUR
TCIAM2151-1G TCIAM2151-100MG
Mirtazapine ≥98.0% (by HPLC)
Mirtazapine
Formula: C₁₇H₁₉N₃
Peso molecolare: 265,36 g/mol
MDL Number: MFCD00865427
CAS Number: 85650-52-8
EINECS: 288-060-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 119.0 to 123.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR