5-Methyl-2-(2-nitroanilino)-3-thiophenecarbonitrile ≥97.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3-Cyano-5-methyl-2-(2-nitroanilino)thiophene, 5-Methyl-2-((2-nitrophenyl)amino)thiophene-3-carbonitrile
M2138-5G
M2138-25G
TCIAM2138-5GEA
111
EUR
TCIAM2138-5G
TCIAM2138-25G
5-Methyl-2-(2-nitroanilino)-3-thiophenecarbonitrile ≥97.0% (by GC)
5-Methyl-2-(2-nitroanilino)-3-thiophenecarbonitrile
Formula:
C₁₂H₉N₃O₂S Peso molecolare: 259,28 g/mol |
MDL Number:
MFCD06408054 CAS Number: 138564-59-7 EINECS: 421-300-1 UN: 3077 ADR: 9,III |
Risultati del test di specificazione
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 109.0 to 115.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...