Stampa…

Milnacipran hydrochloride ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: cis-2-(Aminomethyl)-N,N-diethyl-1-phenylcyclopropanecarboxamide Hydrochloride

M2133-50MG M2133-500MG
TCIAM2133-50MGEA 22 EUR
TCIAM2133-50MG TCIAM2133-500MG
Milnacipran hydrochloride ≥98.0% (by HPLC, titration analysis)
Milnacipran hydrochloride
Formula: C₁₅H₂₂N₂O
Peso molecolare: 282,81 g/mol
Punto di fusione: 181°C
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00901293
CAS Number: 101152-94-7
UN: 2811
ADR: 6.1,III
Merck Index: 14,06194

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR