Methyl-1-adamantanecarboxylate ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Methyl adamantane-1-carboxylate, 1-Adamantanecarboxylic Acid Methyl Ester
M2119-25G
M2119-5G
TCIAM2119-25GEA
208
EUR
TCIAM2119-25G
TCIAM2119-5G
Methyl-1-adamantanecarboxylate ≥98.0% (by GC)
Methyl-1-adamantanecarboxylate
Formula:
C₁₂H₁₈O₂ Peso molecolare: 194,27 g/mol Punto di ebollizione: 79°C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD01838519 CAS Number: 711-01-3 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 35.0 to 39.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...