Stampa…

4-Methoxyphenyl-4-(3-butenyloxy)benzoate ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 4-(3-Butenyloxy)benzoic Acid 4-Methoxyphenyl Ester

M2106-1G M2106-5G
TCIAM2106-1GEA 95 EUR
TCIAM2106-1G TCIAM2106-5G
4-Methoxyphenyl-4-(3-butenyloxy)benzoate ≥98.0% (by GC)
4-Methoxyphenyl-4-(3-butenyloxy)benzoate
Formula: C₁₈H₁₈O₄
Peso molecolare: 298,34 g/mol
MDL Number: MFCD00143337
CAS Number: 76487-56-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0227c442f700bb721b6a8fc37d508cd8e46260c5&vid=0227c442f700bb721b6a8fc37d508cd8e46260c5&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR