4-(4-Methoxybenzyloxy)-1,3-butanediol ≥95.0% (by GC)
Fornitore: TCI
Formula:
C₁₂H₁₈O₄ |
CAS Number:
1820740-81-5 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...