Stampa…

Mexiletine hydrochloride ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1-(2,6-Dimethylphenoxy)propan-2-amine hydrochloride, 1-(2,6-Dimethylphenoxy)-2-propylamine Hydrochloride, Mexiletine HCl

M2040-1G M2040-5G
TCIAM2040-1GEA 17 EUR
TCIAM2040-1G TCIAM2040-5G
Mexiletine hydrochloride ≥98.0% (by HPLC, total nitrogen)
Mexiletine hydrochloride
Formula: C₁₁H₁₇NO
Peso molecolare: 215,72 g/mol
MDL Number: MFCD00216024
CAS Number: 5370-01-4
EINECS: 226-362-1
Merck Index: 14,06169

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 200.0 to 205.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR