Stampa…

Methyl bromodifluoroacetate ≥97.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: Bromodifluoroacetic Acid Methyl Ester

M2020-25G M2020-5G
TCIAM2020-25GEA 188 EUR
TCIAM2020-25G TCIAM2020-5G
Methyl bromodifluoroacetate ≥97.0% (by GC)
Methyl bromodifluoroacetate
Formula: C₃H₃BrF₂O₂
Peso molecolare: 188,96 g/mol
Punto di ebollizione: 96°C
MDL Number: MFCD06248118
CAS Number: 683-98-7
UN: 2924
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c3edf7d59dfbdad128706e3e295d90b781734a4e&vid=c3edf7d59dfbdad128706e3e295d90b781734a4e&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR