Stampa…

2-(Methacryloyloxy)ethyl-2-(trimethylammonio)ethyl phosphate ≥96.0% (by titrimetric analysis)

Fornitore: TCI EUROPE

Synonyms: Phosphoric Acid 2-(Methacryloyloxy)ethyl 2-(Trimethylammonio)ethyl Ester, 2-(Methacryloyloxy)ethyl (2-(trimethylammonio)ethyl) phosphate, 2-Methacryloyloxyethyl Phosphorylcholine

M2005-1G M2005-5G M2005-25G
TCIAM2005-1GEA 39 EUR
TCIAM2005-1G TCIAM2005-5G TCIAM2005-25G
2-(Methacryloyloxy)ethyl-2-(trimethylammonio)ethyl phosphate ≥96.0% (by titrimetric analysis)
2-(Methacryloyloxy)ethyl-2-(trimethylammonio)ethyl phosphate
Formula: C₁₁H₂₂NO₆P
Peso molecolare: 295,27 g/mol
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD11112180
CAS Number: 67881-98-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR