Methyl trans-4-aminocyclohexanecarboxylate hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Fornitore: TCI EUROPE
Synonyms:
trans-4-Aminocyclohexanecarboxylic Acid Methyl Ester Hydrochloride
M1986-1G
M1986-5G
TCIAM1986-1GEA
46
EUR
TCIAM1986-1G
TCIAM1986-5G
Methyl trans-4-aminocyclohexanecarboxylate hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Methyl trans-4-aminocyclohexanecarboxylate hydrochloride
Formula:
C₈H₁₅NO₂ Peso molecolare: 193,67 g/mol |
MDL Number:
MFCD08274538 CAS Number: 61367-07-5 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...