1-Methyl-1-propylpiperidinium bromide ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
Synonyms:
1-Methyl-1-propylpiperidinium Bromide
M1976-5G
M1976-25G
TCIAM1976-5GEA
50
EUR
TCIAM1976-5G
TCIAM1976-25G
1-Methyl-1-propylpiperidinium bromide ≥98.0% (by titrimetric analysis)
1-Methyl-1-propylpiperidinium bromide
Formula:
C₉H₂₀BrN Peso molecolare: 222,17 g/mol |
MDL Number:
MFCD17014743 CAS Number: 88840-42-0 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...