Stampa…

7-Methoxy-4-quinolinol ≥97.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-Hydroxy-7-methoxyquinoline

M1966-1G M1966-5G
TCIAM1966-1GEA 60 EUR
TCIAM1966-1G TCIAM1966-5G
7-Methoxy-4-quinolinol ≥97.0% (by titrimetric analysis)
7-Methoxy-4-quinolinol
Formula: C₁₀H₉NO₂
Peso molecolare: 175,19 g/mol
Punto di fusione: 285°C
MDL Number: MFCD00169015
CAS Number: 82121-05-9

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR