3-Maleimidopropionic acid ≥97.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)propanoic acid, 3-Maleimido-propionic acid, N-(2-Carboxyethyl)maleimide, MPS-Acid, N-Maleoyl-β-alanine
M1962-1G
M1962-200MG
M1962-5G
TCIAM1962-1GEA
156
EUR
TCIAM1962-1G
TCIAM1962-200MG
TCIAM1962-5G
3-Maleimidopropionic acid ≥97.0% (by GC)
3-Maleimidopropionic acid
Formula:
C₇H₇NO₄ Peso molecolare: 169,14 g/mol |
MDL Number:
MFCD00043030 CAS Number: 7423-55-4 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 108.0 to 112.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...