1-Methyl-N-carbobenzoxy-L-glutamate ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
Synonyms:
N-Carbobenzoxy-L-glutamic Acid 1-Methyl Ester, Z-Glu-OMe, 1-Methyl N-Cbz-L-glutamate, (S)-4-(((Benzyloxy)carbonyl)amino)-5-methoxy-5-oxopentanoic acid, N-Cbz-L-glutamic Acid 1-Methyl Ester
M1961-1G
M1961-5G
TCIAM1961-1GEA
53
EUR
TCIAM1961-1G
TCIAM1961-5G
1-Methyl-N-carbobenzoxy-L-glutamate ≥98.0% (by titrimetric analysis)
1-Methyl-N-carbobenzoxy-L-glutamate
Formula:
C₁₄H₁₇NO₆ Peso molecolare: 295,29 g/mol |
MDL Number:
MFCD00083278 CAS Number: 5672-83-3 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 67.0 to 73.0 deg-C |
Specific rotation [a]20/D | -21.0 to -27.0 deg (C=1, EtOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...