Stampa…

ɑ-[2-(Methylamino)ethyl]benzyl alcohol ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-Hydroxy-N-methyl-3-phenylpropylamine, 3-(Methylamino)-1-phenylpropanol, 3-(Methylamino)-1-phenylpropan-1-ol

M1960-25G M1960-5G
TCIAM1960-25GEA 85 EUR
TCIAM1960-25G TCIAM1960-5G
ɑ-[2-(Methylamino)ethyl]benzyl alcohol ≥98.0% (by GC, titration analysis)
alpha-[2-(Methylamino)ethyl]benzyl alcohol
Formula: C₁₀H₁₅NO
Peso molecolare: 165,24 g/mol
Punto di ebollizione: 170°C
MDL Number: MFCD00674078
CAS Number: 42142-52-9
EINECS: 255-679-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 62.0 to 66.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR