Stampa…

Meloxicam ≥98.0% (by HPLC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 4-Hydroxy-2-methyl-N-(5-methylthiazol-2-yl)-2H-benzo[e][1,2]thiazine-3-carboxamide 1,1-dioxide, 4-Hydroxy-2-methyl-N-(5-methyl-2-thiazolyl)-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide

M1959-25G M1959-5G
TCIAM1959-25GEA 56 EUR
TCIAM1959-25G TCIAM1959-5G
Meloxicam ≥98.0% (by HPLC)
Meloxicam
Formula: C₁₄H₁₃N₃O₄S₂
Peso molecolare: 351,40 g/mol
Punto di fusione: 255°C
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00868752
CAS Number: 71125-38-7
UN: 2811
ADR: 6.1,III
Merck Index: 14,05826

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Green powder to crystaline
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR