Stampa…

(R)-1-Methyl-3-pyrrolidinol ≥98.0% (by GC)

Fornitore: TCI
Attenzione

M1937-1G M1937-5G
TCIAM1937-1GEA 53 EUR
TCIAM1937-1G TCIAM1937-5G
(R)-1-Methyl-3-pyrrolidinol ≥98.0% (by GC)
(R)-1-Methyl-3-pyrrolidinol
Formula: C₅H₁₁NO
Punto di ebollizione: 106°C
Flash Pt: 71°C
MDL Number: MFCD03788747
CAS Number: 104641-60-3

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Specific rotation [a]20/D -5.0 to -8.0 deg (C=1, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR