Stampa…

Methyl-2,3,6-tri-O-benzoyl-ɑ-D-galactopyranoside ≥98.0% (by HPLC)

Fornitore: TCI

M1933-25G M1933-250G M1933-5G
TCIAM1933-25GEA 130 EUR
TCIAM1933-25G TCIAM1933-250G TCIAM1933-5G
Methyl-2,3,6-tri-O-benzoyl-ɑ-D-galactopyranoside ≥98.0% (by HPLC)
Methyl-2,3,6-tri-O-benzoyl-alpha-D-galactopyranoside
Formula: C₂₈H₂₆O₉
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD06200842
CAS Number: 3601-36-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Specific rotation [a]20/D +118.0 to +128.0 deg (C=1, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR