2-Methyl-3-(propyldithio)furan ≥97.0% (by GC)
Fornitore: TCI
|
Pericolo
|
Synonyms:
2-Methyl-3-(propyldisulphanyl)furan, 2-Methyl-3-furyl Propyl Disulphide
Formula:
C₈H₁₂OS₂ Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD01729816 CAS Number: 61197-09-9 EINECS: 262-650-3 UN: 2810 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | Light yellow to Brown clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...