Stampa…
Mono(2-acryloyloxyethyl) succinate ≥90.0% (by GC) stabilizzato
Fornitore: TCI EUROPE
Pericolo
Synonyms: 2-(Acryloyloxy)ethyl Hydrogen Succinate, 4-(2-(Acryloyloxy)ethoxy)-4-oxobutanoic acid, Succinic Acid Mono(2-acryloyloxyethyl) Ester
Stabilization: Stabilised with 4-methoxyphenol (MEHQ)
M1824-25G M1824-500G
TCIAM1824-25GEA 17 EUR
TCIAM1824-25G TCIAM1824-500G
Mono(2-acryloyloxyethyl) succinate ≥90.0% (by GC) stabilizzato
Mono(2-acryloyloxyethyl) succinate
Formula: C₉H₁₂O₆
Peso molecolare: 216,19 g/mol
Flash Pt: 178°C
MDL Number: MFCD00274301
CAS Number: 50940-49-3
Visualizza i codici
Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 90.0 %