10-Methylacridinium perchlorate ≥98.0% (by total nitrogen basis)
Fornitore: TCI EUROPE
|
Attenzione
|
M1787-1G
M1787-250MG
TCIAM1787-1GEA
166
EUR
TCIAM1787-1G
TCIAM1787-250MG
10-Methylacridinium perchlorate ≥98.0% (by total nitrogen basis)
10-Methylacridinium perchlorate
Formula:
C₁₄H₁₂ClNO₄ Peso molecolare: 293,70 g/mol |
CAS Number:
26456-05-3 UN: 1479 ADR: 5.1,III |
Risultati del test di specificazione
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...