Stampa…

Mefenamic acid ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 2-(2,3-Xylidino)benzoic acid, 2-((2,3-Dimethylphenyl)amino)benzoic acid, 2-(2,3-Dimethylphenylamino)benzoic acid, N-(2,3-Dimethylphenyl)anthranilic acid

M1782-25G M1782-500G M1782-100G
TCIAM1782-25GEA 30 EUR
TCIAM1782-25G TCIAM1782-500G TCIAM1782-100G
Mefenamic acid ≥98.0% (by titrimetric analysis)
Mefenamic acid
Formula: C₁₅H₁₅NO₂
Peso molecolare: 241,29 g/mol
Punto di fusione: 230°C
MDL Number: MFCD00051721
CAS Number: 61-68-7
EINECS: 200-513-1
Merck Index: 14,05798
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5345375d041702cf922fb762285171bea8fded72&vid=5345375d041702cf922fb762285171bea8fded72&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) min. 98.0 %
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR