Stampa…

2-Methyl-1-naphthyl acetate ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 1-Acetoxy-2-methylnaphthalene, Acetic Acid 2-Methyl-1-naphthyl Ester

M1757-25G M1757-250G
TCIAM1757-25GEA 32 EUR
TCIAM1757-25G TCIAM1757-250G
2-Methyl-1-naphthyl acetate ≥98.0% (by GC)
2-Methyl-1-naphthyl acetate
Formula: C₁₃H₁₂O₂
Peso molecolare: 200,24 g/mol
CAS Number: 5697-02-9

Visualizza i codici

Risultati del test di specificazione

Purity (GC) min. 98.0 %
Solubility in Methanol almost transparency
Melting point 80.0 to 84.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR