4-Methoxyphenyl-3-O-benzyl-4,6-O-benzylidene-β-D-glucopyranoside ≥98.0% (by HPLC)
Fornitore: TCI EUROPE
M1640-1G
M1640-5G
TCIAM1640-1GEA
243
EUR
TCIAM1640-1G
TCIAM1640-5G
4-Methoxyphenyl-3-O-benzyl-4,6-O-benzylidene-β-D-glucopyranoside ≥98.0% (by HPLC)
4-Methoxyphenyl-3-O-benzyl-4,6-O-benzylidene-beta-D-glucopyranoside
Formula:
C₂₇H₂₈O₇ Peso molecolare: 464,51 g/mol Temperatura di stoccaggio: Congelamento |
CAS Number:
303127-81-3 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 204.0 to 211.0 deg-C |
Specific rotation [a]20/D | -17.0 to -24.0 deg (C=1, CHCl3) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...