Stampa…

4-Methoxyphenyl-2,6-di-O-benzyl-β-D-galactopyranoside ≥98.0% (by HPLC)

Fornitore: TCI

M1634-1G M1634-5G
TCIAM1634-1GEA 78 EUR
TCIAM1634-1G TCIAM1634-5G
4-Methoxyphenyl-2,6-di-O-benzyl-β-D-galactopyranoside ≥98.0% (by HPLC)
4-Methoxyphenyl-2,6-di-O-benzyl-beta-D-galactopyranoside
Formula: C₂₇H₃₀O₇
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD06797145
CAS Number: 159922-50-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9868bc9eed2a7ac976e4af966ad7524c3ad6db4&vid=c9868bc9eed2a7ac976e4af966ad7524c3ad6db4&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 109.0 to 113.0 deg-C
Specific rotation [a]20/D 0.0 to -2.0 deg (C=1, CHCl3)
Solubility in Chloroform almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR