Stampa…

4-Methoxyphenyl-2,6-di-O-benzyl-3,4-O-isopropylidene-β-D-galactopyranoside ≥98.0% (by HPLC)

Fornitore: TCI

M1633-1G M1633-5G
TCIAM1633-1GEA 72 EUR
TCIAM1633-1G TCIAM1633-5G
4-Methoxyphenyl-2,6-di-O-benzyl-3,4-O-isopropylidene-β-D-galactopyranoside ≥98.0% (by HPLC)
4-Methoxyphenyl-2,6-di-O-benzyl-3,4-O-isopropylidene-beta-D-galactopyranoside
Formula: C₃₀H₃₄O₇
Peso molecolare: 506.60 g/mol
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD06797144
CAS Number: 159922-68-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dff2a2bcf0e1ee77ae0afc08469bcd44cc10e2a2&vid=dff2a2bcf0e1ee77ae0afc08469bcd44cc10e2a2&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 102.0 to 107.0 deg-C
Specific rotation [a]20/D +13.0 to +16.0 deg (C=1, CHCl3)
Solubility in Chloroform very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR