4-Methoxyphenyl-β-D-glucopyranoside ≥97.0% (by HPLC)
Fornitore: TCI EUROPE
Synonyms:
(2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(4-methoxyphenoxy)tetrahydro-2H-pyran-3,4,5-triol
M1631-25G
M1631-5G
TCIAM1631-25GEA
620
EUR
TCIAM1631-25G
TCIAM1631-5G
4-Methoxyphenyl-β-D-glucopyranoside ≥97.0% (by HPLC)
4-Methoxyphenyl-beta-D-glucopyranoside
Formula:
C₁₃H₁₈O₇ Peso molecolare: 286,28 g/mol Punto di fusione: 176°C Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD06797143 CAS Number: 6032-32-2 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) (CAD) | min. 97.0 area% |
Melting point | 175.0 to 179.0 deg-C |
Specific rotation [a]20/D | -62.0 to -67.0 deg (C=1, H2O) |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...