Stampa…

Methyl-2,3,4-tri-O-acetyl-1-thio-β-L-fucopyranoside ≥98.0% (by HPLC)

Fornitore: TCI

M1626-1G M1626-5G
TCIAM1626-1GEA 144 EUR
TCIAM1626-1G TCIAM1626-5G
Methyl-2,3,4-tri-O-acetyl-1-thio-β-L-fucopyranoside ≥98.0% (by HPLC)
Methyl-2,3,4-tri-O-acetyl-1-thio-beta-L-fucopyranoside
Formula: C₁₃H₂₀O₇S
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00080803
CAS Number: 84635-54-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 144.0 to 148.0 deg-C
Specific rotation [a]20/D 0.0 to +3.0 deg (C=1, CHCl3)
Solubility in Chloroform almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR