Stampa…

4-Methoxyphenyl-3,4-O-isopropylidene-β-D-galactopyranoside ≥98.0% (by HPLC)

Fornitore: TCI EUROPE

M1593-1G M1593-5G
TCIAM1593-1GEA 78 EUR
TCIAM1593-1G TCIAM1593-5G
4-Methoxyphenyl-3,4-O-isopropylidene-β-D-galactopyranoside ≥98.0% (by HPLC)
4-Methoxyphenyl-3,4-O-isopropylidene-beta-D-galactopyranoside
Formula: C₁₆H₂₂O₇
Peso molecolare: 326,35 g/mol
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD06797135
CAS Number: 159922-67-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=721855d10ea05adda79c36a7e662d1bca71ea2d1&vid=721855d10ea05adda79c36a7e662d1bca71ea2d1&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 149.0 to 153.0 deg-C
Specific rotation [a]20/D -17.0 to -20.0 deg (C=1,MeOH)
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR