Stampa…

Morpholinosulphur trifluoride ≥93.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: Morph-DAST

M1573-1G M1573-5G
TCIAM1573-1GEA 25 EUR
TCIAM1573-1G TCIAM1573-5G
Morpholinosulphur trifluoride ≥93.0% (by titrimetric analysis)
Morpholinosulphur trifluoride
Formula: C₄H₈F₃NOS
Peso molecolare: 175,17 g/mol
Punto di ebollizione: 42°C
Flash Pt: 76°C
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00037057
CAS Number: 51010-74-3
UN: 3265
ADR: 8,II

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light orange to Yellow clear liquid
Purity (Neutralization titration) min. 93.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR