1-Adamantyl methacrylate ≥98.0% (by GC) stabilizzato
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
1-Methacryloyloxyadamantane, Methacrylic acid 1-Adamantyl ester
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
M1526-25G
M1526-5G
TCIAM1526-25GEA
267
EUR
TCIAM1526-25G
TCIAM1526-5G
1-Adamantyl methacrylate ≥98.0% (by GC) stabilizzato
1-Adamantyl methacrylate
Formula:
C₁₄H₂₀O₂ Peso molecolare: 220,31 g/mol Punto di ebollizione: 113°C |
MDL Number:
MFCD08703330 CAS Number: 16887-36-8 |
Risultati del test di specificazione
Appearance | White or Colorless to Almost white or Almost colorless powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...