Stampa…

4-Methoxyphenyl-β-D-galactopyranoside ≥98.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: (2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(4-methoxyphenoxy)tetrahydro-2H-pyran-3,4,5-triol

M1481-25G M1481-5G
TCIAM1481-25GEA 291 EUR
TCIAM1481-25G TCIAM1481-5G
4-Methoxyphenyl-β-D-galactopyranoside ≥98.0% (by HPLC)
4-Methoxyphenyl-beta-D-galactopyranoside
Formula: C₁₃H₁₈O₇
Peso molecolare: 286,28 g/mol
MDL Number: MFCD06797126
CAS Number: 3150-20-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) (CAD) min. 98.0 area%
Melting point 157.0 to 162.0 deg-C
Specific rotation [a]20/D -40.0 to -44.0 deg (C=1, MeOH)
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR