11-[4-(4-Butylphenylazo)phenoxy]undecyl methacrylate ≥97.0% (by GC)
Fornitore: TCI
Synonyms:
Methacrylic Acid 11-[4-(4-Butylphenylazo)phenoxy]undecyl Ester
Formula:
C₃₁H₄₄N₂O₃ |
MDL Number:
MFCD06797121 CAS Number: 942230-11-7 |
Risultati del test di specificazione
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 73.0 to 78.0 deg-C |
Solubility in Toluene | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...