Stampa…

Isopropyl 3-mercaptopropionate ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-Mercaptopropionic Acid Isopropyl Ester

M1435-25G M1435-5G
TCIAM1435-25GEA 113 EUR
TCIAM1435-25G TCIAM1435-5G
Isopropyl 3-mercaptopropionate ≥98.0% (by GC)
Isopropyl 3-mercaptopropionate
Formula: C₆H₁₂O₂S
Peso molecolare: 148,22 g/mol
MDL Number: MFCD00040066
CAS Number: 7383-64-4

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR