4,4'-Methylenebis(2-methylcyclohexylamine) (mixture of isomers) ≥99.0% (by GC)
Fornitore: TCI
|
Pericolo
|
Synonyms:
Bis(4-amino-3-methylcyclohexyl)methane, 4,4'-Methylenebis(2-methylcyclohexylamine), 4,4'-Methylenebis(1-amino-2-methylcyclohexane), 4,4'-Diamino-3,3'-dimethyldicyclohexylmethane
Formula:
C₁₅H₃₀N₂ Peso molecolare: 238.42 g/mol |
MDL Number:
MFCD00075535 CAS Number: 6864-37-5 EINECS: 229-962-1 UN: 2927 ADR: 6.1 II |
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 99.0 % (total of isomers) |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...